siRNA Details
Gene Symbol: Ilf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026374.3 | NP_080650.1 | 7, 8 | 500 | ||
GenBank mRNA | AK012364.1 | - | 7, 8 | 487 | ||
AK013858.1 | - | 7, 8 | 546 | |||
AK078003.1 | - | 7, 8 | 487 | |||
AK135449.1 | - | 7, 8 | 575 | |||
AK135539.1 | - | 7, 8 | 487 | |||
AK157129.1 | - | 7, 8 | 555 | |||
BC024718.1 | - | 7, 8 | 500 | 5012365 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
