siRNA Details
Gene Symbol: Rgs8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026380.3 | NP_080656.2 | 7 | 931 | ||
GenBank mRNA | AK044337.1 | - | 7 | 805 | ||
AK142301.1 | - | 7 | 800 | |||
BC065056.1 | - | 7 | 864 | 5705033 | ||
BC094577.1 | - | 6 | 547 | 6852696 |
