siRNA Details
Gene Symbol: Limd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172397.3 | NP_765985.1 | 5 | 398 | ||
GenBank mRNA | AK012581.1 | - | 5 | 372 | ||
AK032430.1 | - | 5 | 399 | |||
AK040486.1 | - | 5 | 399 | |||
AK049809.1 | - | 5 | 368 | |||
AK167349.1 | - | 5 | 368 | |||
AK169829.1 | - | 5 | 369 | |||
AK171794.1 | - | 5 | 677 | |||
AK188791.1 | - | 2 | 97 | |||
BC068130.1 | - | 5 | 342 | 6413206 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
