siRNA Details
Gene Symbol: Snx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026386.1 | NP_080662.1 | 11 | 1142 | ||
GenBank mRNA | AK002692.1 | - | 11 | 1142 | ||
AK005470.1 | - | 11 | 1154 | |||
AK010572.1 | - | 11 | 1139 | |||
AK075929.1 | - | 11 | 1139 | |||
AK145840.1 | - | 11 | 1193 | |||
AK153525.1 | - | 11 | 1139 | |||
BC006960.1 | - | 11 | 1142 | 2812557 | ||
CT010229.1 | - | 11 | 1127 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001106135, XM_006254716, BC166853, CO567837.
Gene Ontology Categories:
