siRNA Details
Gene Symbol: Rmdn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033136.3 | NP_001028308.1 | 11, 12 | 1424 | ||
GenBank mRNA | AK004786.1 | - | 11, 12 | 1350 | ||
AK146299.1 | - | 11, 12 | 1343 | |||
AK151090.1 | - | 11, 12 | 1345 | |||
AK153322.1 | - | 11, 12 | 1345 | |||
AK153369.1 | - | 11, 12 | 1344 | |||
BC055754.1 | - | 11, 12 | 1318 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
