siRNA Details
Gene Symbol: Nmral1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290761.1 | NP_001277690.1 | 5 | 1218 | ||
NM_001290762.1 | NP_001277691.1 | 5 | 1295 | |||
NM_026393.2 | NP_080669.1 | 6 | 1248 | |||
GenBank mRNA | AA166262.1 | - | 1 | 284 | ||
AK012340.1 | - | 5 | 1051 | |||
AK146521.1 | - | 974 | ||||
BC030039.1 | - | 6 | 951 | 1313824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
