siRNA Details
Gene Symbol: Lce1f | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026394.3 | NP_080670.2 | 2 | 668 | ||
GenBank mRNA | AK003518.1 | - | 2 | 669 | ||
BC060283.1 | - | 2 | 652 | |||
BC100541.1 | - | 1 | 636 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
