siRNA Details
Gene Symbol: Wdr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026399.2 | NP_080675.2 | 3, 4 | 472 | ||
GenBank mRNA | AK005790.1 | - | 3, 4 | 412 | ||
AK020654.1 | - | 3, 4 | 433 | |||
AY365434.1 | - | 3, 4 | 472 | |||
BC005587.1 | - | 3 | 711 | |||
BC019369.1 | - | 3, 4 | 395 | 4014928 | ||
BC020047.1 | - | 1314 | ||||
BC034842.1 | - | 3 | 640 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
