siRNA Details
Gene Symbol: 1700021F05Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026411.1 | NP_080687.1 | 2 | 278 | ||
GenBank mRNA | AK006199.1 | - | 2 | 278 | ||
AK007370.1 | - | 2 | 282 | |||
AK008795.1 | - | 2 | 242 | |||
AK163304.1 | - | 2 | 246 | |||
BC013506.1 | - | 2 | 252 | 4224442 |
