siRNA Details
Gene Symbol: Ppp6c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024209.2 | NP_077171.1 | 3, 4 | 342 | ||
GenBank mRNA | AK002764.1 | - | 3, 4 | 304 | ||
AK009104.1 | - | 3, 4 | 322 | |||
BC002223.1 | - | 3, 4 | 367 | 3491111 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_133589, X77236, BC078747.
Gene Ontology Categories:
