siRNA Details
Gene Symbol: Cysrt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026415.3 | NP_080691.2 | 2 | 272 | ||
GenBank mRNA | AK003658.1 | - | 2 | 240 | ||
AK009095.1 | - | 2 | 274 | |||
BC117777.2 | - | 1 | 572 | 40091453 | ||
BC117778.2 | - | 1 | 572 | 40091454 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
