siRNA Details
Gene Symbol: 2310033P09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024210.2 | NP_077172.1 | 2 | 327 | ||
GenBank mRNA | AK009601.1 | - | 2 | 314 | ||
AK012628.1 | - | 2 | 338 | |||
BC002181.1 | - | 2 | 327 | 3487283 |

More Information
Other Species Targeted

Gene Ontology Categories:
