siRNA Details
Gene Symbol: Rprm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023396.5 | NP_075885.1 | 1 | 513 | ||
GenBank mRNA | AB043586.1 | - | 1 | 502 | ||
AK010465.1 | - | 1 | 514 | |||
AK014769.1 | - | 1 | 478 | |||
BC030065.1 | - | 1 | 336 | 1434823 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
