siRNA Details
Gene Symbol: Rprm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023396.5 | NP_075885.1 | 1 | 1081 | ||
GenBank mRNA | AB043586.1 | - | 1 | 1070 | ||
AK010465.1 | - | 1 | 1082 | |||
AK014769.1 | - | 1 | 1046 | |||
BC030065.1 | - | 1 | 904 | 1434823 |

More Information
Other Species Targeted

Gene Ontology Categories:
