siRNA Details
Gene Symbol: Dcxr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026428.2 | NP_080704.2 | 5, 6 | 499 | ||
GenBank mRNA | AK004023.1 | - | 5, 6 | 498 | ||
AK007627.1 | - | 5, 6 | 511 | |||
AK153521.1 | - | 5, 6 | 480 | |||
AK161732.1 | - | 455 | ||||
BC012247.1 | - | 5, 6 | 488 | 4189635 | ||
D89656.1 | - | 5, 6 | 447 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
