siRNA Details
Gene Symbol: 1810043G02Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026431.2 | NP_080707.2 | 6 | 656 | ||
GenBank mRNA | AK003097.1 | - | 6 | 664 | ||
AK004032.1 | - | 6 | 658 | |||
AK075794.1 | - | 6 | 643 | |||
AK147511.1 | - | 2 | 4419 | |||
AK153233.1 | - | 636 | ||||
AK165293.1 | - | 3 | 3457 | |||
BC006038.1 | - | 4 | 527 | |||
BC010330.1 | - | 6 | 627 | 2651637 | ||
BC016605.1 | - | 5 | 1381 | |||
BC029982.1 | - | 6 | 636 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
