siRNA Details
Gene Symbol: Camsap2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081360.1 | NP_001074829.1 | 13 | 2028 | ||
GenBank mRNA | AK028542.1 | - | 11 | 1978 | ||
BC138367.1 | - | 12 | 1604 | 8861387 | ||
BC145258.1 | - | 11 | 1571 | 9053811 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
