siRNA Details
Gene Symbol: Ufm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026435.5 | NP_080711.1 | 4 | 195 | ||
GenBank mRNA | AK007788.1 | - | 4 | 159 | ||
AK008280.1 | - | 4 | 158 | |||
AK011737.1 | - | 4 | 176 | |||
AK089948.1 | - | 4 | 161 | |||
AK148146.1 | - | 4 | 165 | |||
AK166332.1 | - | 4 | 189 | |||
AK167301.1 | - | 4 | 164 | |||
AK167321.1 | - | 4 | 159 | |||
BC051554.1 | - | 3 | 159 | |||
BC061065.1 | - | 4 | 194 | 6594260 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
