siRNA Details
Gene Symbol: Coa6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_174987.4 | NP_778152.1 | 2 | 344 | ||
GenBank mRNA | AK008041.1 | - | 2 | 279 | ||
AK009404.1 | - | 2 | 318 | |||
BC024399.1 | - | 2 | 274 | 5026081 | ||
BC038634.1 | - | 2 | 330 | 1513986 | ||
BC046907.1 | - | 3 | 235 | 6394880 | ||
BU562971.1 | - | 2 | 347 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
