siRNA Details
Gene Symbol: Tmem86a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026436.3 | NP_080712.1 | 3 | 724 | ||
GenBank mRNA | AK007864.1 | - | 3 | 725 | ||
AK075589.1 | - | 3 | 698 | |||
BC056619.1 | - | 3 | 691 | 4987281 |
