siRNA Details
Gene Symbol: Pef1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026441.4 | NP_080717.2 | 4 | 613 | ||
GenBank mRNA | AK007394.1 | - | 4 | 555 | ||
AK013238.1 | - | 4 | 555 | |||
AK046247.1 | - | 4 | 555 | |||
AK075978.1 | - | 4 | 618 | |||
BC019191.1 | - | 4 | 561 | 5029503 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
