siRNA Details
Gene Symbol: 1600012H06Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001083880.1 | NP_001077349.1 | 2 | 694 | ||
NM_001083881.1 | NP_001077350.1 | 2 | 764 | |||
NM_001083882.1 | NP_001077351.1 | 1 | 946 | |||
NM_026451.2 | NP_080727.2 | 2 | 699 | |||
GenBank mRNA | AK005427.1 | - | 2 | 676 | ||
AK017276.1 | - | 1 | 876 | |||
AK028052.1 | - | 2 | 700 | |||
AK076083.1 | - | 2 | 657 | |||
AK076476.1 | - | 2 | 619 | |||
AK083064.1 | - | 1 | 887 | |||
AK150772.1 | - | 2 | 719 | |||
AK167490.1 | - | 2 | 706 | |||
BC027313.1 | - | 2 | 678 | 3710655 | ||
BC043303.1 | - | 1 | 907 | |||
BC043449.1 | - | 1 | 907 | |||
BC054359.1 | - | 2 | 696 | |||
BC071250.1 | - | 2 | 611 |

More Information
Other Species Targeted

Gene Ontology Categories:
