siRNA Details
Gene Symbol: Coq9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026452.2 | NP_080728.1 | 7 | 817 | ||
GenBank mRNA | AK004527.1 | - | 3 | 242 | ||
AK009179.1 | - | 7 | 829 | |||
AK154343.1 | - | 7 | 842 | |||
AK158979.1 | - | 7 | 836 | |||
BC036386.1 | - | 7 | 817 | 1067021 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
