siRNA Details
Gene Symbol: Ppap2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_080555.2 | NP_542122.1 | 2 | 653 | ||
GenBank mRNA | AK011276.1 | - | 2 | 652 | ||
AK076183.1 | - | 2 | 349 | |||
AK159136.1 | - | 2 | 739 | |||
AK160056.1 | - | 2 | 553 | |||
BC005558.1 | - | 2 | 640 | 3964817 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
