siRNA Details
Gene Symbol: Ccdc70 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026459.3 | NP_080735.1 | 2 | 590 | ||
GenBank mRNA | AK006920.1 | - | 1 | 483 | ||
AK007190.1 | - | 2 | 522 | |||
BC049553.1 | - | 2 | 590 | 6773824 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
