siRNA Details
Gene Symbol: Wdr55 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026464.2 | NP_080740.2 | 7 | 1047 | ||
GenBank mRNA | AK017695.1 | - | 7 | 1021 | ||
AK018469.1 | - | 7 | 1028 | |||
AK075958.1 | - | 7 | 1047 | |||
AK146093.1 | - | 7 | 1066 | |||
BC064750.1 | - | 7 | 1006 | |||
BC131932.1 | - | 7 | 1041 | 40130209 | ||
BC131934.1 | - | 7 | 1041 | 40130211 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
