siRNA Details
Gene Symbol: Prorsd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163454.2 | NP_001156926.1 | 3 | 548 | ||
NM_026465.3 | NP_080741.1 | 2 | 457 | |||
GenBank mRNA | AK008574.1 | - | 2 | 458 | ||
AK077280.1 | - | 3 | 546 | |||
AK085965.1 | - | 3 | 525 | |||
BC033930.1 | - | 3 | 521 | 1397170 | ||
BC115900.1 | - | 2 | 458 | 40091622 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
