siRNA Details
Gene Symbol: Atp5g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026468.2 | NP_080744.1 | 4, 5 | 397 | ||
GenBank mRNA | AK002311.1 | - | 4, 5 | 394 | ||
AK007747.1 | - | 4, 5 | 398 | |||
AK012279.1 | - | 4, 5 | 394 | |||
AK075793.1 | - | 4, 5 | 398 | |||
BC006813.1 | - | 3, 4 | 366 | 3155185 | ||
BC081437.1 | - | 4, 5 | 383 | 6410913 | ||
BC106138.1 | - | 3, 4 | 342 | 4983197 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
