siRNA Details
Gene Symbol: Mesdc2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023403.3 | NP_075892.3 | 3 | 783 | ||
GenBank mRNA | AK008735.1 | - | 3 | 500 | ||
AK031949.1 | - | 3 | 526 | |||
AK076760.1 | - | 3 | 523 | |||
AK076773.1 | - | 3 | 518 | |||
BC014742.1 | - | 3 | 504 | 4239248 | ||
BC058612.1 | - | 3437 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
