siRNA Details
Gene Symbol: Spata6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026470.3 | NP_080746.3 | 10 | 1204 | ||
GenBank mRNA | AK005819.1 | - | 5 | 591 | ||
AK006861.1 | - | 5 | 576 | |||
AK051574.1 | - | 10 | 1205 | |||
AK132771.1 | - | 10 | 1178 | |||
AK153098.1 | - | 10 | 1081 | |||
BC028429.1 | - | 5 | 604 | |||
BC040024.1 | - | 5 | 604 |
