siRNA Details
Gene Symbol: Tubb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026473.2 | NP_080749.2 | 1 | 73 | ||
GenBank mRNA | AK009958.1 | - | 1 | 84 | ||
AK144815.1 | - | 1 | 88 | |||
AK151643.1 | - | 1 | 70 | |||
BC008225.1 | - | 1 | 73 | 2631779 |

More Information
Other Species Targeted

Gene Ontology Categories:
