siRNA Details
Gene Symbol: Tomm20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024214.2 | NP_077176.1 | 2, 3 | 358 | ||
GenBank mRNA | AK002902.1 | - | 2, 3 | 279 | ||
AK160168.1 | - | 2, 3 | 261 | |||
AK181461.1 | - | 2, 3 | 77 | |||
AK185977.1 | - | 77 | ||||
BC002087.1 | - | 2, 3 | 267 | 3484983 | ||
BC099449.1 | - | 2, 3 | 265 | 30848526 | ||
DN174518.1 | - | 2, 3 | 358 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_152935, BC078715, D63411, U21871, FQ230650, and others.
Gene Ontology Categories:
