siRNA Details
Gene Symbol: Zcchc10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026479.4 | NP_080755.2 | 2, 3 | 220 | ||
GenBank mRNA | AK010803.1 | - | 2, 3 | 231 | ||
AK020177.1 | - | 2, 3 | 221 | |||
AK027985.1 | - | 2, 3 | 228 | |||
AK169856.1 | - | 2, 3 | 221 | |||
BC025078.1 | - | 2, 3 | 236 | 4504126 | ||
BC087902.1 | - | 2, 3 | 220 | 30505854 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
