siRNA Details
Gene Symbol: Ooep | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026480.3 | NP_080756.1 | 3 | 469 | ||
GenBank mRNA | AB283026.1 | - | 3 | 409 | ||
AK010266.1 | - | 3 | 464 | |||
AK010809.1 | - | 3 | 470 | |||
AK133554.1 | - | 3 | 434 | |||
AK135950.1 | - | 3 | 435 | |||
AK136199.1 | - | 435 | ||||
AK145510.1 | - | 3 | 435 | |||
BC062245.1 | - | 3 | 420 | 3470921 | ||
DQ238107.1 | - | 3 | 470 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
