siRNA Details
Gene Symbol: Tppp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026481.3 | NP_080757.1 | 2, 3 | 346 | ||
GenBank mRNA | AK009771.1 | - | 2, 3 | 310 | ||
AK012437.1 | - | 2, 3 | 314 | |||
AK155018.1 | - | 2, 3 | 317 | |||
AK160507.1 | - | 2, 3 | 317 | |||
BC010788.1 | - | 2, 3 | 303 | 4216285 | ||
BC116622.1 | - | 1, 2 | 138 | |||
BF469439.1 | - | 2, 3 | 346 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
