siRNA Details
Gene Symbol: Pdzd9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040136.2 | NP_001035226.1 | 4 | 745 | ||
NM_001040137.2 | NP_001035227.1 | 4 | 515 | |||
GenBank mRNA | AK005921.1 | - | 4 | 746 | ||
AK006727.1 | - | 4 | 649 | |||
AK015115.1 | - | 4 | 516 | |||
AK015578.1 | - | 4 | 516 | |||
AK017019.1 | - | 4 | 653 | |||
BC147551.1 | - | 4 | 741 | 9056542 | ||
BC147563.1 | - | 4 | 741 | 9056553 | ||
BC172089.1 | - | 3 | 551 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
