siRNA Details
Gene Symbol: Mrps11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026498.2 | NP_080774.2 | 5 | 664 | ||
GenBank mRNA | AB049945.1 | - | 5 | 388 | ||
AK002993.1 | - | 5 | 501 | |||
AK152027.1 | - | 5 | 455 | |||
AK160207.1 | - | 5 | 444 | |||
AK166241.1 | - | 5 | 452 | |||
BC005701.1 | - | 5 | 406 | |||
BC028279.1 | - | 5 | 456 | 5370270 |

More Information
Other Species Targeted

Gene Ontology Categories:
