siRNA Details
Gene Symbol: Srsf6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026499.4 | NP_080775.3 | 3 | 503 | ||
GenBank mRNA | AK004831.1 | - | 3 | 484 | ||
AK012605.1 | - | 3 | 251 | |||
AK051608.1 | - | 1 | 656 | |||
AK133872.1 | - | 3 | 484 | |||
AK146293.1 | - | 3 | 481 | |||
AK159519.1 | - | 3 | 481 | |||
AK171709.1 | - | 2 | 1767 | |||
AK179065.1 | - | 2 | 132 | |||
AK179529.1 | - | 1 | 88 | |||
AK188277.1 | - | 2 | 112 | |||
AK212807.1 | - | 1 | 88 | |||
BC012039.1 | - | 3 | 490 | 2648111 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001014185, BC087121, FM060375.
Gene Ontology Categories:
