siRNA Details
Gene Symbol: 1110004E09Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026502.2 | NP_080778.2 | 2 | 400 | ||
GenBank mRNA | AK003413.1 | - | 2 | 291 | ||
AK018510.1 | - | 2 | 371 | |||
AK045905.1 | - | 2 | 379 | |||
BC019533.1 | - | 2 | 364 | 4191729 | ||
CX236612.1 | - | 2 | 400 |
