siRNA Details
Gene Symbol: Sdhaf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026503.3 | NP_080779.2 | 2, 3 | 252 | ||
GenBank mRNA | AK003789.1 | - | 2, 3 | 210 | ||
AK004301.1 | - | 2, 3 | 232 | |||
AK007909.1 | - | 2, 3 | 245 | |||
AK015824.1 | - | 2, 3 | 253 | |||
BC028981.1 | - | 2, 3 | 217 | 5367409 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
