siRNA Details
Gene Symbol: Apopt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163388.1 | NP_001156860.1 | 2 | 227 | ||
NM_026511.2 | NP_080787.1 | 2 | 227 | |||
GenBank mRNA | AK002873.1 | - | 2 | 229 | ||
AK005403.1 | - | 2 | 215 | |||
AK010493.1 | - | 2 | 217 | |||
AK012648.1 | - | 2 | 225 | |||
AK029408.1 | - | 2 | 203 | |||
AK148446.1 | - | 2 | 167 | |||
AK160910.1 | - | 2 | 211 | |||
BC071262.1 | - | 2 | 227 | 6509633 |
