siRNA Details
Gene Symbol: 2810417H13Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026515.2 | NP_080791.2 | 2, 3 | 224 | ||
GenBank mRNA | AK011090.1 | - | 2, 3 | 230 | ||
AK011645.1 | - | 2, 3 | 187 | |||
AK011835.1 | - | 2, 3 | 184 | |||
AK012140.1 | - | 2, 3 | 194 | |||
AK017673.1 | - | 2, 3 | 187 | |||
AY496943.1 | - | 2, 3 | 118 | |||
BC021413.1 | - | 2, 3 | 170 | 5006272 | ||
BC096393.1 | - | 2, 3 | 160 | 1397865 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
