siRNA Details
Gene Symbol: Fam122a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026520.3 | NP_080796.1 | 1 | 712 | ||
GenBank mRNA | AK005219.1 | - | 1 | 702 | ||
AK010777.1 | - | 1 | 723 | |||
AK011741.1 | - | 1 | 696 | |||
AK033646.1 | - | 1 | 712 | |||
AK077654.1 | - | 1 | 694 | |||
AK172089.1 | - | 1 | 697 | |||
BC028637.1 | - | 1 | 680 | 1248203 | ||
BC083062.1 | - | 1 | 702 | 5361132 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001014029, BC078922.
Gene Ontology Categories:
