siRNA Details
Gene Symbol: Nmb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291280.1 | NP_001278209.1 | 2 | 382 | ||
NM_026523.4 | NP_080799.1 | 2 | 382 | |||
GenBank mRNA | AA671747.1 | - | 2 | 353 | ||
AK011929.1 | - | 2 | 339 | |||
AK014077.1 | - | 2 | 361 | |||
BC028490.1 | - | 2 | 317 | 1366735 | ||
CB058776.1 | - | 2 | 394 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
