siRNA Details
Gene Symbol: Zfp593 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024215.2 | NP_077177.2 | 3 | 643 | ||
GenBank mRNA | AK014081.1 | - | 2 | 687 | ||
AK133585.1 | - | 3 | 641 | |||
AK156931.1 | - | 3 | 578 | |||
AK214591.1 | - | 1 | 36 | |||
BC003465.1 | - | 3 | 580 | 2648747 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
