siRNA Details
Gene Symbol: Nutf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026532.3 | NP_080808.1 | 4 | 335 | ||
GenBank mRNA | AK002461.1 | - | 4 | 317 | ||
AK012500.1 | - | 4 | 317 | |||
AK020512.1 | - | 4 | 334 | |||
AK028410.1 | - | 4 | 325 | |||
AK051049.1 | - | 4 | 317 | |||
AK152212.1 | - | 4 | 345 | |||
AK168026.1 | - | 4 | 317 | |||
BC003955.1 | - | 4 | 335 | 3487899 | ||
BC083165.1 | - | 4 | 389 | 6437914 | ||
BC086773.1 | - | 4 | 302 | 6828661 |

More Information
Other Species Targeted

Gene Ontology Categories:
