siRNA Details
Gene Symbol: MDFI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001300804.1 | NP_001287733.1 | 6 | 1359 | ||
NM_001300805.1 | NP_001287734.1 | 4 | 1039 | |||
NM_001300806.1 | NP_001287735.1 | 4 | 1308 | |||
NM_005586.3 | NP_005577.1 | 5 | 1222 | |||
XM_005249117.3 | XP_005249174.1 | 5 | 1225 | |||
XM_011514625.2 | XP_011512927.1 | 6 | 1510 | |||
XM_011514626.2 | XP_011512928.1 | 5 | 1333 | |||
XM_017010867.1 | XP_016866356.1 | 4 | 1881 | |||
GenBank mRNA | AK290735.1 | - | 4 | 1306 | ||
BC007836.2 | - | 5 | 1171 | 4110841 | ||
U78313.1 | - | 5 | 1164 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
