siRNA Details
Gene Symbol: Rps13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026533.3 | NP_080809.1 | 4 | 282 | ||
GenBank mRNA | AK012479.1 | - | 4 | 227 | ||
AK018703.1 | - | 4 | 226 | |||
AK076060.1 | - | 4 | 226 | |||
AK088237.1 | - | 3 | 859 | |||
AK151043.1 | - | 4 | 227 | |||
AK151671.1 | - | 4 | 228 | |||
AK161798.1 | - | 3 | 825 | |||
AK166691.1 | - | 4 | 227 | |||
AK177445.1 | - | 225 | ||||
AV498110.1 | - | 3 | 189 | |||
BC011192.1 | - | 4 | 225 | 4016679 | ||
BC085269.1 | - | 2 | 931 | |||
BC090397.1 | - | 4 | 209 | 5150054 | ||
BC106137.1 | - | 4 | 210 | 4990874 | ||
BC125602.1 | - | 4 | 226 | 40130117 | ||
BC125604.1 | - | 4 | 226 | 40130119 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
