siRNA Details
Gene Symbol: Serpina12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026535.2 | NP_080811.1 | 2 | 805 | ||
GenBank mRNA | AK014346.1 | - | 2 | 747 | ||
AK014589.1 | - | 2 | 805 | |||
AY326419.1 | - | 1 | 638 | |||
BC062143.1 | - | 2 | 759 | 30287005 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
