siRNA Details
Gene Symbol: Atp5s | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026536.1 | NP_080812.1 | 4 | 411 | ||
GenBank mRNA | AK003725.1 | - | 4 | 411 | ||
AK009141.1 | - | 5 | 523 | |||
BC027442.1 | - | 4 | 393 | 4015414 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
